Assay Details
Target Gene Details
Entrez Gene ID: | 100534612 |
Gene Name: | C1QTNF3-AMACR readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.5:33986986-34124528 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1QTNF3-AMACR | NR_037951.1 | |||
AB209261.1 | BAD92498.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830255 | Chr.5:33929507 - 34109160 on Build GRCh38 | Loss | C1QTNF3 C1QTNF3-AMACR RXFP3 SLC45A2 AMACR |
nsv519240 | Chr.5:33950598 - 34749311 on Build GRCh38 | Gain | C1QTNF3 LOC105374753 C1QTNF3-AMACR LOC646652 SLC45A2 AMACR RAI14 |
nsv522715 | Chr.5:33319157 - 34037875 on Build GRCh38 | Gain | C1QTNF3 C1QTNF3-AMACR RXFP3 TARS SLC45A2 AMACR ADAMTS12 |