Assay Details
Target Gene Details
Entrez Gene ID: | 241 |
Gene Name: | arachidonate 5-lipoxygenase activating protein |
Gene Aliases: |
FLAP |
Location: |
Chr.13:30713478-30764428 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALOX5AP | NM_001204406.1 | NP_001191335.1 | ||
NM_001629.3 | NP_001620.2 | |||
XM_017020522.1 | XP_016876011.1 | |||
AK314143.1 | ||||
BC018538.1 | AAH18538.1 | |||
BG569867.1 | ||||
BQ003388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv984 | Chr.13:30747467 - 30777530 on Build GRCh38 | Insertion | ALOX5AP |
nsv832574 | Chr.13:30657196 - 30822408 on Build GRCh38 | Gain | USPL1 LINC00398 ALOX5AP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |