Assay Details
Target Gene Details
Entrez Gene ID: | 102724020 |
Gene Name: | uncharacterized LOC102724020 |
Gene Aliases: |
- |
Location: |
Chr.12:10015352-10030606 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC102724020 | NR_120484.1 | |||
BU570676.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1374n100 | Chr.12:9968689 - 10026775 on Build GRCh38 | Gain | LOC102724020 CLEC12B CLEC12A CLEC1B |