Assay Details
Target Gene Details
Entrez Gene ID: | 7166 |
Gene Name: | tryptophan hydroxylase 1 |
Gene Aliases: |
TPRH, TRPH |
Location: |
Chr.11:18020537-18040788 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TPH1 | NM_004179.2 | NP_004170.1 | ||
BC106739.1 | AAI06740.1 | |||
BC106740.2 | ||||
BP423529.1 | ||||
X52836.1 | CAA37018.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625482 | Chr.11:18029957 - 18032997 on Build GRCh38 | Loss | TPH1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |