Assay Details
Target Gene Details
Entrez Gene ID: | 51043 |
Gene Name: | zinc finger and BTB domain containing 7B |
Gene Aliases: |
CKROX, THPOK, ZBTB15, ZFP-67, ZFP67, ZNF857B, c-KROX, hcKROX |
Location: |
Chr.1:155002628-155018525 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZBTB7B | NM_001252406.2 | NP_001239335.1 | ||
NM_001256455.1 | NP_001243384.1 | |||
NR_045515.1 | ||||
NR_046206.1 | ||||
NR_049765.1 | ||||
XM_006711349.2 | XP_006711412.2 | |||
XM_006711353.1 | XP_006711416.1 | |||
XM_006711354.1 | XP_006711417.1 | |||
XM_006711359.2 | XP_006711422.1 | |||
AK291619.1 | ||||
AK304762.1 | ||||
BC012070.1 | AAH12070.1 | |||
BF510901.1 | ||||
BM801567.1 | ||||
CR749303.1 | CAH18158.1 | |||
CV572166.1 | ||||
DA970906.1 | ||||
DB241444.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion | ZBTB7B ADAM15 LOC100505666 DPM3 EFNA4 EFNA1 DCST1 LENEP LOC101928120 MIR4258 DCST2 PMVK EFNA3 PBXIP1 SHC1 FLAD1 CKS1B PYGO2 SLC50A1 |
nsv952212 | Chr.1:154998225 - 155087624 on Build GRCh38 | Deletion | ZBTB7B ADAM15 EFNA3 LOC100505666 EFNA4 DCST1 DCST2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |