Assay Details
Target Gene Details
Entrez Gene ID: | 203427 |
Gene Name: | solute carrier family 25 member 43 |
Gene Aliases: |
- |
Location: |
Chr.X:119399295-119454478 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A43 | NM_145305.2 | NP_660348.2 | ||
AK091304.1 | ||||
AK094254.1 | BAC04318.1 | |||
AK292035.1 | ||||
AK303932.1 | ||||
BC019584.1 | AAH19584.1 | |||
BC071871.1 | AAH71871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv518389 | Chr.X:119301240 - 119460459 on Build GRCh38 | Loss | SLC25A43 LOC101928336 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |