Assay Details
Target Gene Details
Entrez Gene ID: | 84457 |
Gene Name: | phytanoyl-CoA 2-hydroxylase interacting protein like |
Gene Aliases: |
- |
Location: |
Chr.10:59174811-59247774 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHYHIPL | NM_001143774.1 | NP_001137246.1 | ||
NM_032439.3 | NP_115815.2 | |||
XM_011540275.2 | XP_011538577.1 | |||
XM_011540276.2 | XP_011538578.1 | |||
XM_017016782.1 | XP_016872271.1 | |||
XM_017016783.1 | XP_016872272.1 | |||
AK054956.1 | BAB70832.1 | |||
AL365474.1 | CAB97028.1 | |||
AL834339.1 | CAD39006.1 | |||
BC011268.2 | AAH11268.2 | |||
CR749429.1 | CAH18267.1 | |||
DB455338.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831888 | Chr.10:59183895 - 59378493 on Build GRCh38 | Gain | FAM13C PHYHIPL |