Assay Details
Target Gene Details
Entrez Gene ID: | 7581 |
Gene Name: | zinc finger protein 33A |
Gene Aliases: |
KOX2, KOX31, KOX5, NF11A, ZNF11, ZNF11A, ZNF33, ZZAPK |
Location: |
Chr.10:38010483-38067834 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF33A | NR_136709.1 | |||
NR_136710.1 | ||||
AK027057.1 | BAB15642.1 | |||
BC030511.1 | AAH30511.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1040704 | Chr.10:37651096 - 38162989 on Build GRCh38 | Gain | ZNF33BP1 ZNF248 ZNF37A ZNF33A ZNF25 |
esv3576438 | Chr.10:38002320 - 38607542 on Build GRCh38 | Gain | HSD17B7P2 SEPT7P9 LINC00999 ZNF37A ZNF33A LOC100129055 |
nsv550419 | Chr.10:38022849 - 38212726 on Build GRCh38 | Gain | ZNF37A ZNF33A LOC100129055 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |