Assay Details
Target Gene Details
Entrez Gene ID: | 8879 |
Gene Name: | sphingosine-1-phosphate lyase 1 |
Gene Aliases: |
S1PL, SPL |
Location: |
Chr.10:70815920-70881189 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SGPL1 | NM_003901.3 | NP_003892.2 | ||
XM_005270263.1 | XP_005270320.1 | |||
XM_006718053.1 | XP_006718116.1 | |||
XM_011540316.2 | XP_011538618.1 | |||
XM_011540317.1 | XP_011538619.1 | |||
XM_011540318.1 | XP_011538620.1 | |||
AB033078.1 | ||||
AF144638.1 | AAD44755.1 | |||
AJ011304.2 | CAA09590.2 | |||
AK314615.1 | ||||
BC052991.1 | AAH52991.1 | |||
BP314104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1043245 | Chr.10:70290210 - 70915897 on Build GRCh38 | Loss | ADAMTS14 EIF4EBP2 PRF1 PCBD1 NODAL TBATA LRRC20 PALD1 SGPL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |