Assay Details
Target Gene Details
Entrez Gene ID: | 85313 |
Gene Name: | peptidylprolyl isomerase like 4 |
Gene Aliases: |
HDCME13P |
Location: |
Chr.6:149504495-149546102 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPIL4 | NM_139126.3 | NP_624311.1 | ||
AF068298.1 | AAF65187.1 | |||
AF308286.1 | AAG48254.1 | |||
AF357880.1 | AAM63961.1 | |||
AK315388.1 | ||||
BC016984.1 | AAH16984.1 | |||
BC020986.1 | AAH20986.1 | |||
BC064134.1 | AAH64134.1 | |||
BX537536.1 | CAD97776.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |