Assay Details
Target Gene Details
Entrez Gene ID: | 5188 |
Gene Name: | glutamyl-tRNA amidotransferase subunit B |
Gene Aliases: |
HSPC199, PET112, PET112L |
Location: |
Chr.4:151669773-151761023 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GATB | NM_004564.2 | NP_004555.1 | ||
XM_005263063.3 | XP_005263120.1 | |||
AB019410.2 | ||||
AF026851.1 | AAD08640.1 | |||
AF151033.1 | AAF36119.1 | |||
AK222887.1 | BAD96607.1 | |||
AK312957.1 | ||||
BC130348.1 | ||||
BC136547.1 | ||||
BX537555.1 | CAH56162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv4551 | Chr.4:151661465 - 151706215 on Build GRCh38 | Deletion | FAM160A1 GATB |
nsv1028814 | Chr.4:150874356 - 152285445 on Build GRCh38 | Gain | SH3D19 RPS3A LOC100505685 LOC100996286 SNORD73A LRBA PRSS48 FAM160A1 GATB |
nsv1026511 | Chr.4:151672230 - 151690763 on Build GRCh38 | Gain | GATB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |