Assay Details
Target Gene Details
Entrez Gene ID: | 127933 |
Gene Name: | U2AF homology motif (UHM) kinase 1 |
Gene Aliases: |
KIS, KIST, P-CIP2 |
Location: |
Chr.1:162497174-162529629 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UHMK1 | NM_001184763.1 | NP_001171692.1 | ||
NM_144624.2 | NP_653225.2 | |||
NM_175866.4 | NP_787062.1 | |||
AJ536197.1 | CAD60192.1 | |||
AK058195.1 | ||||
AK292369.1 | ||||
AL834136.1 | CAI46199.1 | |||
BC014917.1 | AAH14917.1 | |||
BC026046.1 | AAH26046.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758973 | Chr.1:162094678 - 162533566 on Build GRCh38 | Gain | C1orf111 SH2D1B MIR4654 MIR556 NOS1AP UHMK1 C1orf226 |
nsv516482 | Chr.1:162138329 - 162905405 on Build GRCh38 | Gain+Loss | C1orf111 SH2D1B CCDC190 MIR4654 MIR556 NOS1AP UAP1 DDR2 HSD17B7 UHMK1 C1orf226 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |