Assay Details
Target Gene Details
Entrez Gene ID: | 221477 |
Gene Name: | chromosome 6 open reading frame 89 |
Gene Aliases: |
BRAP |
Location: |
Chr.6:36834886-36928964 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C6orf89 | XM_017010434.1 | XP_016865923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5262 | Chr.6:36831553 - 36858434 on Build GRCh38 | Deletion | CPNE5 PPIL1 C6orf89 |
nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain | MIR3925 PPIL1 C6orf89 PIM1 MDGA1 SRSF3 GLO1 DNAH8 LOC107986531 LOC100505530 GLP1R PI16 FGD2 CMTR1 MIR4462 PANDAR ZFAND3 CPNE5 LOC100131047 BTBD9 RAB44 CCDC167 MTCH1 TMEM217 RNF8 TBC1D22B CDKN1A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |