Assay Details
Target Gene Details
Entrez Gene ID: | 27190 |
Gene Name: | interleukin 17B |
Gene Aliases: |
IL-17B, IL-20, NIRF, ZCYTO7 |
Location: |
Chr.5:149374267-149404202 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL17B | NM_001317987.1 | NP_001304916.1 | ||
XM_011537622.2 | XP_011535924.1 | |||
XM_017009347.1 | XP_016864836.1 | |||
XM_017009348.1 | XP_016864837.1 | |||
BM826445.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv599948 | Chr.5:149346599 - 149413198 on Build GRCh38 | Loss | IL17B PCYOX1L GRPEL2 GRPEL2-AS1 CARMN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |