Assay Details
Target Gene Details
Entrez Gene ID: | 8974 |
Gene Name: | prolyl 4-hydroxylase subunit alpha 2 |
Gene Aliases: |
- |
Location: |
Chr.5:132190147-132227863 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
P4HA2 | NM_001017973.1 | 15 | 2220 | NP_001017973.1 |
NM_001017974.1 | 15 | 1748 | NP_001017974.1 | |
NM_001142598.1 | 16 | 1884 | NP_001136070.1 | |
NM_001142599.1 | 16 | 1890 | NP_001136071.1 | |
NM_004199.2 | 15 | 2226 | NP_004190.1 | |
XM_005272116.4 | 15 | 1754 | XP_005272173.1 | |
XM_005272117.4 | 16 | 1912 | XP_005272174.1 | |
XM_005272118.4 | 16 | 2041 | XP_005272175.1 | |
XM_005272119.4 | 16 | 2231 | XP_005272176.1 | |
XM_005272120.4 | 16 | 2011 | XP_005272177.1 | |
XM_006714728.3 | 16 | 2288 | XP_006714791.1 | |
XM_006714729.3 | 16 | 1906 | XP_006714792.1 | |
XM_006714730.3 | 16 | 2282 | XP_006714793.1 | |
XM_011543705.2 | 13 | 1503 | XP_011542007.1 | |
XM_017010011.1 | 16 | 2005 | XP_016865500.1 | |
XM_017010012.1 | 14 | 1571 | XP_016865501.1 | |
AY358970.1 | 15 | 1822 | AAQ89329.1 | |
BC017062.1 | 15 | 1849 | ||
BC035813.1 | 15 | 1717 | AAH35813.1 | |
BX647924.1 | 15 | 1740 | CAI46066.1 | |
U90441.1 | 15 | 1849 | AAB71339.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs71810391] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |