Assay Details
Target Gene Details
Entrez Gene ID: | 51302 |
Gene Name: | cytochrome P450 family 39 subfamily A member 1 |
Gene Aliases: |
- |
Location: |
Chr.6:46549580-46652849 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYP39A1 | NM_001278738.1 | 12 | 1659 | NP_001265667.1 |
NM_001278739.1 | 11 | 1544 | NP_001265668.1 | |
NM_016593.4 | 12 | 1719 | NP_057677.2 | |
XM_011514658.1 | 10 | 1509 | XP_011512960.1 | |
XM_017010924.1 | 11 | 1586 | XP_016866413.1 | |
XM_017010925.1 | 9 | 1401 | XP_016866414.1 | |
AF237982.1 | 12 | 1515 | AAF63329.1 | |
AK292263.1 | 12 | 1651 | ||
AK301604.1 | ||||
AK307986.1 | 11 | 1208 | ||
BC010358.1 | 12 | 1679 | AAH10358.1 | |
BG199773.1 | 1 | 614 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |