Assay Details
Target Gene Details
Entrez Gene ID: | 686 |
Gene Name: | biotinidase |
Gene Aliases: |
- |
Location: |
Chr.3:15601352-15653709 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BTD |
Target Gene Details
Entrez Gene ID: | 26061 |
Gene Name: | 2-hydroxyacyl-CoA lyase 1 |
Gene Aliases: |
2-HPCL, HPCL, HPCL2, PHYH2 |
Location: |
Chr.3:15560704-15601852 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HACL1 | NM_001284413.1 | 1 | 419 | NP_001271342.1 |
NM_001284415.1 | 1 | 419 | NP_001271344.1 | |
NM_001284416.1 | 1 | 419 | NP_001271345.1 | |
NM_012260.3 | 1 | 419 | NP_036392.2 | |
NR_104315.1 | 1 | 419 | ||
AF161397.1 | 1 | 39 | AAF28957.1 | |
AJ131753.1 | 1 | 128 | CAB60200.1 | |
AK056977.1 | 1 | 136 | ||
AK301546.1 | 1 | 136 | ||
AK301990.1 | 1 | 154 | ||
AK302086.1 | 1 | 136 | ||
BC001627.1 | 1 | 137 | AAH01627.1 | |
BC007440.2 | 1 | 124 | AAH07440.1 | |
DA609383.1 | 1 | 136 | ||
DB049608.1 | 1 | 190 | ||
DB241165.1 | 1 | 419 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic non-DGV Variation |