Assay Details
Target Gene Details
Entrez Gene ID: | 883 |
Gene Name: | kynurenine aminotransferase 1 |
Gene Aliases: |
CCBL1, GTK, KAT1, KATI |
Location: |
Chr.9:128832942-128882551 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KYAT1 | NM_001122671.1 | 6 | 698 | NP_001116143.1 |
NM_001122672.1 | 5 | 554 | NP_001116144.1 | |
NM_001287390.1 | 8 | 1095 | NP_001274319.1 | |
NM_004059.4 | 6 | 704 | NP_004050.3 | |
NR_109829.1 | 6 | 704 | ||
XM_006717311.3 | 7 | 890 | XP_006717374.1 | |
XM_011519165.2 | 8 | 1066 | XP_011517467.1 | |
XM_011519167.2 | 6 | 670 | XP_011517469.2 | |
XM_011519168.1 | 5 | 529 | XP_011517470.1 | |
XM_011519169.1 | 6 | 798 | XP_011517471.1 | |
XM_011519170.2 | 6 | 1166 | XP_011517472.1 | |
XM_017015260.1 | 7 | 850 | XP_016870749.1 | |
XM_017015261.1 | 8 | 1072 | XP_016870750.1 | |
XM_017015262.1 | 8 | 1561 | XP_016870751.1 | |
XM_017015263.1 | 8 | 976 | XP_016870752.1 | |
XM_017015264.1 | 7 | 883 | XP_016870753.1 | |
XM_017015265.1 | 6 | 1260 | XP_016870754.1 | |
XM_017015266.1 | 7 | 891 | XP_016870755.1 | |
XM_017015267.1 | 6 | 712 | XP_016870756.1 | |
XM_017015268.1 | 7 | 874 | XP_016870757.1 | |
AK094505.1 | 6 | 1247 | ||
AK291178.1 | 6 | 692 | ||
AK297995.1 | 8 | 951 | ||
AK314427.1 | 6 | 574 | ||
AU123740.1 | 6 | 682 | ||
BC033685.1 | 5 | 408 | AAH33685.1 | |
BI755163.1 | 5 | 408 | ||
EL733697.1 | 4 | 369 | ||
X82224.1 | 5 | 519 | CAA57702.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6724 | Chr.9:128823206 - 128858415 on Build GRCh38 | Insertion | SPOUT1 KYAT1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |