Assay Details
Target Gene Details
Entrez Gene ID: | 9796 |
Gene Name: | phytanoyl-CoA 2-hydroxylase interacting protein |
Gene Aliases: |
DYRK1AP3, PAHX-AP, PAHXAP1 |
Location: |
Chr.8:22219703-22232338 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHYHIP | NM_001099335.1 | 6 | 1894 | NP_001092805.1 |
NM_014759.3 | 5 | 1772 | NP_055574.3 | |
XM_006716416.1 | XP_006716479.1 | |||
XM_017014102.1 | 3 | 2037 | XP_016869591.1 | |
AK098743.1 | 6 | 1517 | ||
AK294372.1 | 4 | 1224 | ||
BC034034.1 | 5 | 1525 | AAH34034.1 | |
BC035940.1 | 5 | 1525 | AAH35940.1 | |
D87463.1 | 6 | 1603 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470205 | Chr.8:22191277 - 22256052 on Build GRCh38 | Loss | LOC100507071 PHYHIP MIR320A POLR3D BMP1 |
dgv12054n54 | Chr.8:22045805 - 22251098 on Build GRCh38 | Loss | DMTN FAM160B2 FGF17 HR PHYHIP LGI3 REEP4 MIR320A POLR3D NUDT18 SFTPC BMP1 |
nsv831265 | Chr.8:22199688 - 22366918 on Build GRCh38 | Loss | PIWIL2 LOC100507071 PHYHIP MIR320A POLR3D BMP1 |
nsv831264 | Chr.8:22093700 - 22328040 on Build GRCh38 | Gain+Loss | PIWIL2 LOC100507071 FAM160B2 HR PHYHIP LGI3 REEP4 MIR320A POLR3D NUDT18 SFTPC BMP1 |
esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss | LOC107986876 PIWIL2 TNFRSF10B FGF17 LOC389641 CHMP7 REEP4 PPP3CC TNFRSF10C SFTPC TNFRSF10D BIN3 DOK2 LOC100507071 DMTN LOC286059 SORBS3 PDLIM2 GFRA2 LOC107984124 LOC254896 PEBP4 SLC39A14 NUDT18 BMP1 BIN3-IT1 LOC101929237 C8orf58 TNFRSF10A HR NPM2 LOC102467222 LOC101929172 XPO7 EGR3 FAM160B2 CCAR2 RHOBTB2 PHYHIP LGI3 MIR320A LOC286114 POLR3D |
More Information
Additional Information:
Set Membership: |
Intragenic Exonic DGV Variation Validated |