Assay Details
Target Gene Details
Entrez Gene ID: | 7054 |
Gene Name: | tyrosine hydroxylase |
Gene Aliases: |
DYT14, DYT5b, TYH |
Location: |
Chr.11:2163929-2174081 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TH | NM_000360.3 | NP_000351.2 | ||
NM_199292.2 | NP_954986.2 | |||
NM_199293.2 | NP_954987.2 | |||
XM_011520335.2 | XP_011518637.1 | |||
AA447751.1 | ||||
BC104967.1 | AAI04968.1 | |||
BC143611.1 | ||||
BC143614.1 | ||||
BM718799.1 | ||||
DQ677336.1 | ||||
DQ677337.1 | ||||
M17589.1 | AAA61179.1 | |||
X05290.1 | CAA28908.1 | |||
Y00414.1 | CAA68472.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv553070 | Chr.11:2154812 - 2177188 on Build GRCh38 | Loss | INS-IGF2 INS TH MIR4686 |
nsv469927 | Chr.11:2165105 - 2201683 on Build GRCh38 | Loss | TH MIR4686 |
nsv553068 | Chr.11:2096173 - 2174751 on Build GRCh38 | Loss | INS-IGF2 IGF2 INS MIR483 TH MIR4686 IGF2-AS |
nsv951284 | Chr.11:2083671 - 2218670 on Build GRCh38 | Deletion | INS-IGF2 IGF2 INS MIR483 TH MIR4686 IGF2-AS |
nsv553047 | Chr.11:1733170 - 2418537 on Build GRCh38 | Gain | INS-IGF2 SNORD131 IGF2 CD81-AS1 TNNI2 HOTS TSSC4 LSP1 TRPM5 INS MIR675 MIR483 LINC01219 C11orf21 TNNT3 LINC01150 IFITM10 H19 SYT8 MIR4686 IGF2-AS ASCL2 CD81 TSPAN32 MRPL23 MRPL23-AS1 MIR7847 MIR4298 CTSD TH |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |