Assay Details
Target Gene Details
Entrez Gene ID: | 221785 |
Gene Name: | zinc finger and SCAN domain containing 25 |
Gene Aliases: |
ZNF498 |
Location: |
Chr.7:99616600-99738689 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZSCAN25 | NM_145115.2 | 8 | 1421 | NP_660090.2 |
XM_005250194.2 | 6 | 1193 | XP_005250251.1 | |
XM_011515905.2 | 8 | 1732 | XP_011514207.1 | |
XM_011515906.2 | 9 | 1839 | XP_011514208.1 | |
XM_011515907.2 | 7 | 1676 | XP_011514209.1 | |
XM_011515908.2 | 8 | 1746 | XP_011514210.1 | |
XM_011515909.2 | XP_011514211.1 | |||
XM_011515910.2 | XP_011514212.1 | |||
XM_017011824.1 | 8 | 1468 | XP_016867313.1 | |
XM_017011825.1 | 8 | 1773 | XP_016867314.1 | |
XM_017011826.1 | 9 | 1450 | XP_016867315.1 | |
XM_017011827.1 | XP_016867316.1 | |||
AK057030.1 | 8 | 1398 | ||
AK160369.1 | 3 | 1993 | BAD18712.1 | |
BC069644.1 | 5 | 1086 | AAH69644.2 | |
BC074902.2 | 2 | 412 | ||
BC074903.2 | 2 | 412 | ||
BC089402.1 | 5 | 963 | AAH89402.1 | |
BC114573.1 | 3 | 860 | ||
BC114941.1 | 5 | 1080 | ||
BX640720.1 | CAE45839.1 | |||
BX648158.1 | 7 | 2804 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |