Assay Details
Target Gene Details
Entrez Gene ID: | 220965 |
Gene Name: | family with sequence similarity 13 member C |
Gene Aliases: |
FAM13C1 |
Location: |
Chr.10:59245933-59363186 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM13C | NM_001001971.2 | 10 | 1212 | NP_001001971.1 |
NM_001143773.1 | 13 | 1677 | NP_001137245.1 | |
NM_001166698.1 | 13 | 1588 | NP_001160170.1 | |
NM_198215.3 | 12 | 1506 | NP_937858.2 | |
XM_005269618.4 | 12 | 1503 | XP_005269675.2 | |
XM_005269619.4 | 10 | 1209 | XP_005269676.2 | |
XM_006717702.3 | 14 | 1507 | XP_006717765.1 | |
XM_006717703.3 | 14 | 1689 | XP_006717766.1 | |
XM_011539490.2 | 13 | 1562 | XP_011537792.1 | |
XM_011539491.2 | 14 | 1608 | XP_011537793.1 | |
XM_017015885.1 | 13 | 1420 | XP_016871374.1 | |
XM_017015886.1 | 13 | 1674 | XP_016871375.1 | |
XM_017015887.1 | 13 | 1424 | XP_016871376.1 | |
XM_017015888.1 | 14 | 1686 | XP_016871377.1 | |
XM_017015889.1 | 14 | 1504 | XP_016871378.1 | |
XM_017015890.1 | 14 | 1743 | XP_016871379.1 | |
XM_017015891.1 | 11 | 1383 | XP_016871380.1 | |
XM_017015892.1 | 11 | 1380 | XP_016871381.1 | |
XM_017015893.1 | 13 | 1484 | XP_016871382.1 | |
AK289796.1 | 12 | 1432 | ||
AK294805.1 | 12 | 1421 | ||
AK299924.1 | 13 | 1677 | ||
AK316542.1 | 13 | 1584 | ||
BC036453.1 | 12 | 1464 | AAH36453.1 | |
BC064431.1 | 10 | 1212 | AAH64431.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831888 | Chr.10:59183895 - 59378493 on Build GRCh38 | Gain | PHYHIPL FAM13C |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |