Assay Details
Target Gene Details
Entrez Gene ID: | 68070 |
Gene Name: | PDZ domain containing 2 |
Gene Aliases: |
4930537L06Rik, A930022H17Rik, Gm82, Pdzk3, mKIAA0300 |
Location: |
Chr.15:12357054-12739973 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdzd2 | NM_001081064.1 | NP_001074533.1 | ||
AK159017.1 | BAE34770.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |