Assay Details
Target Gene Details
Entrez Gene ID: | 14229 |
Gene Name: | FK506 binding protein 5 |
Gene Aliases: |
D17Ertd592e, Dit1, FKBP-5, FKBP51 |
Location: |
Chr.17:28398753-28486149 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fkbp5 | NM_010220.4 | 11 | 1829 | NP_034350.1 |
AK169795.1 | 11 | 1770 | BAE41371.1 | |
BC015260.1 | 11 | 1768 | AAH15260.1 | |
U16959.1 | 11 | 1829 | AAA86983.1 | |
U36220.1 | 11 | 1757 | AAA89162.1 |
More Information
Set Membership: |
Intragenic Exonic |