Assay Details
Target Gene Details
Entrez Gene ID: | 13177 |
Gene Name: | enoyl-Coenzyme A delta isomerase 1 |
Gene Aliases: |
Dci, eci |
Location: |
Chr.17:24426683-24439316 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eci1 | NM_010023.4 | 7 | 792 | NP_034153.2 |
AK004838.1 | 7 | 793 | BAB23605.1 | |
AK029481.1 | 7 | 793 | BAC26469.1 | |
AK077386.1 | 7 | 783 | ||
BC022712.1 | 7 | 776 | AAH22712.1 | |
BC054444.1 | 7 | 779 | AAH54444.1 | |
Z14049.1 | 7 | 772 | CAA78417.1 |
More Information
Set Membership: |
Intragenic Exonic |