Assay Details
Target Gene Details
Entrez Gene ID: | 71726 |
Gene Name: | single-strand selective monofunctional uracil DNA glycosylase |
Gene Aliases: |
1200013B09Rik, A930006H09Rik, C85220 |
Location: |
Chr.15:103153290-103168895 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smug1 | NM_027885.3 | 4 | 649 | NP_082161.2 |
AK004735.1 | 3 | 385 | BAB23517.1 | |
AK020817.1 | ||||
AK045411.1 | 2 | 309 | ||
AK172510.1 | 3 | 382 | BAE43042.1 | |
BC050253.1 | 7 | 925 | AAH50253.1 | |
BC062960.1 | 4 | 649 | AAH62960.1 |
More Information
Set Membership: |
Intragenic Exonic |