Assay Details
Target Gene Details
Entrez Gene ID: | 71147 |
Gene Name: | 3-oxoacyl-ACP synthase, mitochondrial |
Gene Aliases: |
4933425A18Rik, C80494 |
Location: |
Chr.14:16238659-16249808 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Oxsm | NM_027695.3 | 2 | 667 | NP_081971.1 |
AK016905.1 | 3 | 621 | BAB30490.1 | |
AK034243.1 | 2 | 374 | ||
AK083617.1 | 2 | 668 | BAC38969.1 | |
AK084829.1 | 2 | 378 | BAC39286.1 | |
BC118051.1 | 1 | 347 | ||
BC119793.1 | 1 | 347 |