Assay Details
Target Gene Details
Entrez Gene ID: | 69906 |
Gene Name: | solute carrier family 25, member 32 |
Gene Aliases: |
2610043O12Rik, Mftc |
Location: |
Chr.15:39094191-39112768 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc25a32 | NM_172402.3 | 7 | 1350 | NP_765990.2 |
AK011759.1 | 1 | 267 | ||
AK031187.1 | 7 | 1155 | BAC27295.1 | |
AK145178.1 | 7 | 1310 | BAE26277.1 | |
AK167501.1 | 7 | 1160 | BAE39577.1 | |
AK170643.1 | 7 | 1319 | BAE41931.1 | |
AK172029.1 | 7 | 1318 | BAE42785.1 | |
BC031874.1 | 7 | 1284 | AAH31874.1 |
More Information
Set Membership: |
Intragenic Exonic |