Assay Details
Target Gene Details
Entrez Gene ID: | 68070 |
Gene Name: | PDZ domain containing 2 |
Gene Aliases: |
4930537L06Rik, A930022H17Rik, Gm82, Pdzk3, mKIAA0300 |
Location: |
Chr.15:12357054-12739973 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdzd2 | NM_001081064.1 | 24 | 9097 | NP_001074533.1 |
AK122240.1 | 6 | 4766 | BAC65522.1 | |
AK136542.1 | 3 | 1222 | BAE23036.1 |
More Information
Set Membership: |
Intragenic Exonic |