Assay Details
Target Gene Details
Entrez Gene ID: | 13116 |
Gene Name: | cytochrome P450, family 46, subfamily a, polypeptide 1 |
Gene Aliases: |
Cyp46 |
Location: |
Chr.12:108334261-108362248 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp46a1 | NM_010010.1 | 15 | 1638 | NP_034140.1 |
AF094479.1 | 15 | 1638 | AAD41243.1 | |
AK140740.1 | 15 | 1643 | BAE24462.1 | |
BC018307.1 | 15 | 1661 | AAH18307.1 |
More Information
Set Membership: |
Intragenic Exonic |