Assay Details
Target Gene Details
Entrez Gene ID: | 14232 |
Gene Name: | FK506 binding protein 8 |
Gene Aliases: |
38kDa, FKBP-38, FKBP-8, Fkbp38 |
Location: |
Chr.8:70527743-70535328 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fkbp8 | NM_001111066.1 | 9 | 1553 | NP_001104536.1 |
NM_001199631.1 | 9 | 1649 | NP_001186560.1 | |
NM_010223.2 | 9 | 1550 | NP_034353.2 | |
AF030635.1 | 8 | 1320 | AAB86422.1 | |
AK088739.1 | 9 | 1488 | BAC40541.1 | |
AK146122.1 | 9 | 1548 | BAE26916.1 | |
AK152198.1 | 9 | 1549 | BAE31027.1 | |
AK153000.1 | 9 | 1542 | ||
AK165281.1 | 9 | 1546 | BAE38118.1 | |
AK167663.1 | 9 | 1536 | BAE39713.1 | |
AK178628.1 | 1 | 278 | ||
AK188492.1 | 1 | 271 | ||
AK189244.1 | 1 | 281 | ||
AK190969.1 | 1 | 252 | ||
AK192821.1 | 1 | 226 | ||
AK205574.1 | 1 | 248 | ||
AK207414.1 | 1 | 231 | ||
AK208969.1 | 1 | 220 | ||
AK212129.1 | 1 | 247 | ||
AK213138.1 | 1 | 256 | ||
AK214360.1 | 1 | 237 | ||
AK218022.1 | 2 | 328 | ||
AY278608.1 | 9 | 1509 | AAQ84562.1 | |
BC003739.1 | 8 | 1469 | AAH03739.1 | |
BC027808.1 | 9 | 1503 | AAH27808.1 |
More Information
Set Membership: |
Intragenic Exonic |