Assay Details
Target Gene Details
Entrez Gene ID: | 71183 |
Gene Name: | C-type lectin domain family 12, member B |
Gene Aliases: |
4933425B16Rik |
Location: |
Chr.6:129375513-129427634 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clec12b |
Target Gene Details
Entrez Gene ID: | 56760 |
Gene Name: | C-type lectin domain family 1, member b |
Gene Aliases: |
1810061I13Rik, Clec-2, Clec2 |
Location: |
Chr.6:129397297-129405413 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clec1b | NM_001204239.1 | 1 | 256 | NP_001191168.1 |
NM_001204253.1 | 1 | 256 | NP_001191182.1 | |
NM_019985.3 | 1 | 256 | NP_064369.1 | |
AF201457.1 | AAF36831.1 | |||
AK007925.1 | 1 | 228 | ||
BC064054.1 | 1 | 256 | AAH64054.1 | |
EF070191.1 |