Assay Details
Target Gene Details
Entrez Gene ID: | 70225 |
Gene Name: | peptidylprolyl isomerase (cyclophilin)-like 3 |
Gene Aliases: |
2310076N22Rik, 2510026K04Rik, Cyp10l |
Location: |
Chr.1:58430993-58445527 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppil3 | NM_001285826.1 | 7 | 1256 | NP_001272755.1 |
NM_001285827.1 | 7 | 921 | NP_001272756.1 | |
NM_027351.3 | 7 | 735 | NP_081627.1 | |
AF315803.1 | 7 | 869 | AAO32944.1 | |
AK010207.1 | 7 | 739 | BAB26768.1 | |
BB687332.1 | 2 | 227 | ||
BC060993.1 | 7 | 703 | AAH60993.1 | |
BC061645.1 | 7 | 1199 | AAH61645.1 | |
DV043111.1 |
More Information
Set Membership: |
Intragenic Exonic |