Assay Details
Target Gene Details
Entrez Gene ID: | 13074 |
Gene Name: | cytochrome P450, family 17, subfamily a, polypeptide 1 |
Gene Aliases: |
Cyp17, p450c17 |
Location: |
Chr.19:46667165-46673000 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp17a1 | NM_007809.3 | 7 | 1348 | NP_031835.3 |
AK132830.1 | 7 | 1242 | ||
AK134376.1 | 7 | 1242 | BAE22120.1 | |
BC064793.1 | 7 | 1322 | AAH64793.1 | |
M64863.1 | 7 | 1221 | AAA39877.1 |