Assay Details
Target Gene Details
Entrez Gene ID: | 64385 |
Gene Name: | cytochrome P450, family 4, subfamily f, polypeptide 14 |
Gene Aliases: |
1300014O15Rik, cyp4f-14 |
Location: |
Chr.17:32905070-32917348 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp4f14 | NM_001204333.1 | NP_001191262.1 | ||
NM_001204334.1 | NP_001191263.1 | |||
NM_001204335.1 | NP_001191264.1 | |||
NM_001204336.1 | NP_001191265.1 | |||
NM_022434.2 | NP_071879.1 | |||
AB037540.1 | BAB12563.1 | |||
AB037541.1 | BAB12564.1 | |||
AF233644.1 | AAK15010.1 | |||
AI047403.1 | ||||
AK005007.1 | BAB23740.1 | |||
AK018676.1 | BAB31338.1 | |||
BC011228.1 | AAH11228.1 | |||
BC094016.1 | AAH94016.1 | |||
BY705215.1 | ||||
CX200266.1 |
More Information
Set Membership: |
Intragenic Exonic |