Assay Details
Target Gene Details
Entrez Gene ID: | 11429 |
Gene Name: | aconitase 2, mitochondrial |
Gene Aliases: |
Aco-2, Aco3, D10Wsu183e |
Location: |
Chr.15:81872463-81915137 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aco2 | NM_080633.2 | NP_542364.1 | ||
AK143917.1 | BAE25602.1 | |||
AK144207.1 | BAE25770.1 | |||
AK145511.1 | BAE26479.1 | |||
AK150027.1 | BAE29252.1 | |||
AK150954.1 | ||||
AK165411.1 | BAE38169.1 | |||
AK191827.1 | ||||
AK198314.1 | ||||
AK198988.1 | ||||
AK199021.1 | ||||
AK204717.1 | ||||
AK206733.1 | ||||
BC004645.1 | AAH04645.1 | |||
BC094462.1 | AAH94462.1 | |||
CA750686.1 |
More Information
Set Membership: |
Intragenic Non-exonic |