Assay Details
Target Gene Details
Entrez Gene ID: | 22355 |
Gene Name: | vasoactive intestinal peptide receptor 2 |
Gene Aliases: |
VPAC2, VPAC2R, Vip2 |
Location: |
Chr.12:116077726-116159604 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vipr2 | NM_009511.2 | 6 | 644 | NP_033537.1 |
AK038743.1 | 6 | 645 | ||
BC138572.1 | 6 | 542 | ||
D28132.1 | 6 | 576 | BAA05674.1 |
More Information
Set Membership: |
Intragenic Exonic |