Assay Details
Target Gene Details
Entrez Gene ID: | 69123 |
Gene Name: | enoyl-Coenzyme A delta isomerase 3 |
Gene Aliases: |
1810022C23Rik, Kseg2 |
Location: |
Chr.13:34946614-34963884 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eci3 |