Assay Details
Target Gene Details
Entrez Gene ID: | 112407 |
Gene Name: | egl-9 family hypoxia-inducible factor 3 |
Gene Aliases: |
2610021G09Rik, AI505553, AI648162, Hif-p4h-3, Phd3, SM-20 |
Location: |
Chr.12:54178981-54203874 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Egln3 | NM_028133.2 | NP_082409.2 | ||
AF421882.1 | AAL17824.1 | |||
AJ310548.1 | CAC42517.1 | |||
AK011502.1 | ||||
AK044787.1 | BAC32092.1 | |||
AK165972.1 | BAE38492.1 | |||
AK170732.1 | BAE41988.1 | |||
BC044926.1 | AAH44926.1 | |||
BC058278.1 | AAH58278.1 | |||
BC069893.1 | AAH69893.1 |