Assay Details
Target Gene Details
Entrez Gene ID: | 30795 |
Gene Name: | FK506 binding protein 3 |
Gene Aliases: |
25kDa, FKBP-3, FKBP25 |
Location: |
Chr.12:65062432-65073938 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fkbp3 | NM_013902.4 | NP_038930.1 | ||
AF110812.1 | AAD20598.1 | |||
AF135595.1 | AAD25097.1 | |||
AK012346.1 | ||||
AK139213.1 | 2 | 4616 | ||
AK145648.1 | BAE26564.1 | |||
AK150802.1 | BAE29865.1 | |||
AK168641.1 | BAE40500.1 | |||
BC002122.1 | AAH02122.1 | |||
U62545.1 | AAB05769.1 |
More Information
Set Membership: |
Intragenic Non-exonic |