Assay Details
Target Gene Details
Entrez Gene ID: | 107817 |
Gene Name: | jumonji domain containing 6 |
Gene Aliases: |
5730436I23Rik, D11Ertd195e, PSR, PtdSerR, Ptdsr, mKIAA0585 |
Location: |
Chr.11:116837432-116843449 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Jmjd6 | NM_033398.2 | NP_203971.2 | ||
AF304118.1 | AAG27719.1 | |||
AK122317.1 | 3 | 2836 | BAC65599.1 | |
BC056629.1 | AAH56629.1 | |||
BC117737.1 | ||||
BP769727.1 | ||||
EF527404.1 | ||||
EF527405.1 |
More Information
Set Membership: |
Intragenic Non-exonic |