Assay Details
Target Gene Details
Entrez Gene ID: | 104112 |
Gene Name: | ATP citrate lyase |
Gene Aliases: |
A730098H14Rik, AW538652 |
Location: |
Chr.11:100476352-100528000 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acly | NM_001199296.1 | 6 | 806 | NP_001186225.1 |
NM_134037.3 | 6 | 806 | NP_598798.1 | |
AF332051.1 | 5 | 601 | AAK56080.1 | |
AF332052.1 | 5 | 601 | AAK56081.1 | |
AK132755.1 | 6 | 697 | BAE21336.1 | |
AK135528.1 | 6 | 673 | ||
AK149660.1 | 6 | 785 | BAE29010.1 | |
AK162364.1 | 6 | 677 | BAE36874.1 | |
AK169703.1 | 6 | 654 | BAE41315.1 | |
BC056378.1 | 6 | 707 | AAH56378.1 |
More Information
Set Membership: |
Intragenic Exonic |