Assay Details
Target Gene Details
Entrez Gene ID: | 104112 |
Gene Name: | ATP citrate lyase |
Gene Aliases: |
A730098H14Rik, AW538652 |
Location: |
Chr.11:100476352-100528000 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acly | NM_001199296.1 | 22 | 2668 | NP_001186225.1 |
NM_134037.3 | 21 | 2638 | NP_598798.1 | |
AF332051.1 | 20 | 2433 | AAK56080.1 | |
AF332052.1 | 20 | 2433 | AAK56081.1 | |
AK043434.1 | 5 | 564 | ||
AK132755.1 | 22 | 2559 | BAE21336.1 | |
AK135528.1 | 22 | 2536 | ||
AK136812.1 | 5 | 476 | BAE23135.1 | |
AK149660.1 | 22 | 2647 | BAE29010.1 | |
AK169703.1 | 21 | 2486 | BAE41315.1 | |
BC021502.1 | 8 | 914 | AAH21502.1 | |
BC056378.1 | 21 | 2539 | AAH56378.1 |
More Information
Set Membership: |
Intragenic Exonic |