Assay Details
Target Gene Details
Entrez Gene ID: | 216343 |
Gene Name: | tryptophan hydroxylase 2 |
Gene Aliases: |
AU043594, Ntph |
Location: |
Chr.10:115078641-115185022 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tph2 | NM_173391.3 | 11 | 1497 | NP_775567.2 |
AY090565.1 | 11 | 1497 | AAM08923.1 | |
BC120514.1 | 11 | 1488 | ||
BC125430.1 | 11 | 1488 |