Assay Details
Target Gene Details
Entrez Gene ID: | 234724 |
Gene Name: | tyrosine aminotransferase |
Gene Aliases: |
- |
Location: |
Chr.8:109990436-109999804 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tat | NM_146214.3 | NP_666326.1 | ||
AK090244.1 | BAC41146.1 | |||
AK149383.1 | BAE28844.1 | |||
AK149526.1 | BAE28939.1 | |||
BC023949.1 | AAH23949.1 | |||
BC024120.1 | AAH24120.1 | |||
BC024264.1 | AAH24264.1 | |||
BC025934.1 | AAH25934.1 | |||
BC028821.1 | AAH28821.1 | |||
BC030728.1 | AAH30728.1 | |||
BC030729.1 | AAH30729.1 | |||
BC037526.1 | AAH37526.1 |
More Information
Set Membership: |
Intragenic Non-exonic |