Assay Details
Target Gene Details
Entrez Gene ID: | 21823 |
Gene Name: | tyrosine hydroxylase |
Gene Aliases: |
- |
Location: |
Chr.7:142892776-142899966 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Th | NM_009377.1 | 12 | 1274 | NP_033403.1 |
M69200.1 | 12 | 1274 | AAA40434.1 |