Assay Details
Target Gene Details
Entrez Gene ID: | 21990 |
Gene Name: | tryptophan hydroxylase 1 |
Gene Aliases: |
Tph |
Location: |
Chr.7:46644641-46672537 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tph1 | NM_001136084.2 | NP_001129556.1 | ||
NM_009414.3 | NP_033440.1 | |||
AK146134.1 | BAE26925.1 | |||
AK146171.1 | BAE26950.1 | |||
BC072582.1 | AAH72582.1 | |||
J04758.1 | AAA63401.1 |