Assay Details
Target Gene Details
Entrez Gene ID: | 74438 |
Gene Name: | clavesin 1 |
Gene Aliases: |
4933402J24Rik, Clvl1, Rlbp1l1 |
Location: |
Chr.4:9144756-9451691 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clvs1 | NM_028940.2 | 3 | 819 | NP_083216.1 |
AK016622.1 | 3 | 743 | BAB30342.1 | |
AK032549.1 | 3 | 808 | ||
AK083810.1 | 3 | 817 | ||
AK084350.1 | 3 | 815 | BAC39166.1 | |
AK136083.1 | 2 | 354 | BAE22811.1 | |
BC062923.1 | 3 | 774 | AAH62923.1 | |
BC100556.1 | 1 | 39 | AAI00557.1 |
More Information
Set Membership: |
Intragenic Exonic |