Assay Details
Target Gene Details
Entrez Gene ID: | 19892 |
Gene Name: | retinal pigment epithelium 65 |
Gene Aliases: |
65kDa, A930029L06Rik, LCA2, Mord1, RP20, rd12 |
Location: |
Chr.3:159599075-159625307 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rpe65 | NM_029987.2 | NP_084263.2 | ||
AF410461.1 | AAL01119.1 | |||
AK020906.1 | BAB32248.1 | |||
BC130028.1 | ||||
CJ133618.1 |
More Information
Set Membership: |
Intragenic Non-exonic |