Assay Details
Target Gene Details
Entrez Gene ID: | 241593 |
Gene Name: | protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, retrogene 1 |
Gene Aliases: |
OTTMUSG00000014964, Pin1-ps1, Pin1l |
Location: |
Chr.2:104713887-104716362 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pin1rt1 | NM_001033768.2 | 1 | 145 | NP_001028940.1 |
AK145372.1 | 1 | 146 | BAE26396.1 |
More Information
Set Membership: |
Intragenic Exonic |